Merck
  • Genetic Depletion of Class I Odorant Receptors Impacts Perception of Carboxylic Acids.
Genetic Depletion of Class I Odorant Receptors Impacts Perception of Carboxylic Acids.
Current biology : CB (2019-08-06)
Annika Cichy, Ami Shah, Adam Dewan, Sarah Kaye, Thomas Bozza
ABSTRACT
The mammalian main olfactory pathway detects myriad volatile chemicals using >1,000 odorant receptor (OR) genes, which are organized into two phylogenetically distinct classes (class I and class II). An important question is how these evolutionarily conserved classes contribute to odor perception. Here, we report functional inactivation of a large number of class I ORs in mice via identification and deletion of a local cis-acting enhancer in the class I gene cluster. This manipulation reduced expression of half of the 131 intact class I genes. The resulting class I-depleted mice exhibited a significant reduction in the number of glomeruli responding to carboxylic acids-chemicals associated with microbial action and body odors. These mice also exhibit a change in odor perception marked by a selective loss of behavioral aversion to these compounds. Together, our data demonstrate that class I ORs play a critical role in representing a class of biologically relevant chemosignals.
Sigma-Aldrich
2-Methylbutyric acid, 98%