Merck
  • Studies on alkaline phosphatase. Phosphorylation of calf-intestinal alkaline phosphatase by 32P-labelled pyrophosphate.
Studies on alkaline phosphatase. Phosphorylation of calf-intestinal alkaline phosphatase by 32P-labelled pyrophosphate.
The Biochemical journal (1968-03-01)
H N Fernley, S Bisaz
ABSTRACT
1. A purified preparation of alkaline phosphatase from calf-intestinal mucosa was phosphorylated by (32)P-labelled PP(i) at a serine residue on the enzyme. Under the conditions employed, up to 0.15mum-labelled sites were obtained from 1mum-[(32)P]PP(i). 2. The phosphorylated enzyme was labile, the rate of dephosphorylation being similar to the overall rate of substrate hydrolysis. 3. A stopped-flow technique was used to determine the number of phosphomonoesterase active sites, which agreed with the number of (32)P-labelled sites. 4. It is concluded that calf-intestinal alkaline phosphatase is both a phosphomonoesterase and a pyrophosphatase.
Sigma-Aldrich
Phosphatase, Acid from potato, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein