Merck
  • Depolarization-induced suppression of a glycinergic synapse in the superior olivary complex by endocannabinoids.
Depolarization-induced suppression of a glycinergic synapse in the superior olivary complex by endocannabinoids.
Journal of neurochemistry (2013-07-19)
Barbara Trattner, Sarah Berner, Benedikt Grothe, Lars Kunz
ABSTRACT
The neuronal endocannabinoid system is known to depress synaptic inputs retrogradely in an activity-dependent manner. This mechanism has been generally described for excitatory glutamatergic and inhibitory GABAergic synapses. Here, we report that neurones in the auditory brainstem of the Mongolian gerbil (Meriones unguiculatus) retrogradely regulate the strength of their inputs via the endocannabinoid system. By means of whole-cell patch-clamp recordings, we found that retrograde endocannabinoid signalling attenuates both glycinergic and glutamatergic post-synaptic currents in the same types of neurones. Accordingly, we detected the cannabinoid receptor 1 in excitatory and inhibitory pre-synapses as well as the endocannabinoid-synthesising enzymes (diacylglycerol lipase α/β, DAGLα/β) post-synaptically through immunohistochemical stainings. Our study was performed with animals aged 10-15 days, that is, in the time window around the onset of hearing. Therefore, we suggest that retrograde endocannabinoid signalling has a role in adapting inputs during the functionally important switch from spontaneously generated to sound-related signals.
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, clone 3C10.2, Chemicon®, from mouse