Merck
  • A lipophilic cation protects crops against fungal pathogens by multiple modes of action.
A lipophilic cation protects crops against fungal pathogens by multiple modes of action.
Nature communications (2020-04-02)
Gero Steinberg, Martin Schuster, Sarah J Gurr, Tina A Schrader, Michael Schrader, Mark Wood, Andy Early, Sreedhar Kilaru
ABSTRACT
The emerging resistance of crop pathogens to fungicides poses a challenge to food security and compels discovery of new antifungal compounds. Here, we show that mono-alkyl lipophilic cations (MALCs) inhibit oxidative phosphorylation by affecting NADH oxidation in the plant pathogens Zymoseptoria tritici, Ustilago maydis and Magnaporthe oryzae. One of these MALCs, consisting of a dimethylsulfonium moiety and a long alkyl chain (C18-SMe2+), also induces production of reactive oxygen species at the level of respiratory complex I, thus triggering fungal apoptosis. In addition, C18-SMe2+ activates innate plant defense. This multiple activity effectively protects cereals against Septoria tritici blotch and rice blast disease. C18-SMe2+ has low toxicity in Daphnia magna, and is not mutagenic or phytotoxic. Thus, MALCs hold potential as effective and non-toxic crop fungicides.
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Hexyltrimethylammonium bromide, ≥98.0% (AT)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, ≥98%
Supelco
Dodin, PESTANAL®, analytical standard
Sigma-Aldrich
Trimethyloctadecylammonium bromide, 98%
Sigma-Aldrich
Sodium monododecyl phosphate, AldrichCPR
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)