Merck
Curcumin recognizes a unique binding site of tubulin.
Journal of medicinal chemistry (2011-08-13)
Soumyananda Chakraborti, Lalita Das, Neha Kapoor, Amlan Das, Vishnu Dwivedi, Asim Poddar, Gopal Chakraborti, Mark Janik, Gautam Basu, Dulal Panda, Pinak Chakrabarti, Avadhesha Surolia, Bhabatarak Bhattacharyya
ABSTRACT
Although curcumin is known for its anticarcinogenic properties, the exact mechanism of its action or the identity of the target receptor is not completely understood. Studies on a series of curcumin analogues, synthesized to investigate their tubulin binding affinities and tubulin self-assembly inhibition, showed that: (i) curcumin acts as a bifunctional ligand, (ii) analogues with substitution at the diketone and acetylation of the terminal phenolic groups of curcumin are less effective, (iii) a benzylidiene derivative, compound 7, is more effective than curcumin in inhibiting tubulin self-assembly. Cell-based studies also showed compound 7 to be more effective than curcumin. Using fluorescence spectroscopy we show that curcumin binds tubulin 32 Å away from the colchicine-binding site. Docking studies also suggests that the curcumin-binding site to be close to the vinblastine-binding site. Structure-activity studies suggest that the tridented nature of compound 7 is responsible for its higher affinity for tubulin compared to curcumin.
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
trans-Cinnamic acid, ≥99%