Merck
  • Berberine inhibits the Warburg effect through TET3/miR-145/HK2 pathways in ovarian cancer cells.
Berberine inhibits the Warburg effect through TET3/miR-145/HK2 pathways in ovarian cancer cells.
Journal of Cancer (2021-01-05)
Jie Li, Yuliang Zou, Meili Pei, Yun Zhang, Yu Jiang
ABSTRACT
Background: Berberine, as an alkaloid, has a significant antitumor effect, but its mechanism in tumor metabolism, especially the Warburg effect has not been elucidated. Objectives: To study the molecular mechanism of berberine regulating the Warburg effect in ovarian cancer cells. Methods: Treatment by berberine in SKOV3 and 3AO cells or inhibited by miR-145 inhibitor transfection in berberine-treated cells to examine the changes in HK2 expression, glucose consumption and lactate production. The methylation status in the promoter region of pre-miR-145 gene was examined by bisulfite sequencing. Dual-luciferase reporter assay was conducted to verify the direct binding of miR-145 to HK2. Finally, the expression of TET3 in ovarian cancer was investigated by quantitative real-time PCR and immunohistochemistry. Results: We found berberine inhibited the Warburg effect by up-regulating miR-145, miR-145 targeted HK2 directly. Berberine increased the expression of miR-145 by promoting the expression of TET3 and reducing the methylation level of the promoter region of miR-145 precursor gene. We further found that TET3 expression was negatively correlated with clinical stage and pathological grade. Conclusions: Our results revealed berberine increased the TET3-mediated demethylation and promoted the suppression of miR-145 on HK2 to antagonize the Warburg effect of ovarian cancer cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human TET3 (1)