Merck
  • Up-regulation of lipolysis genes and increased production of AMP-activated protein kinase protein in the skeletal muscle of rats after resistance training.
Up-regulation of lipolysis genes and increased production of AMP-activated protein kinase protein in the skeletal muscle of rats after resistance training.
Journal of exercise rehabilitation (2016-07-16)
Jae-Heung An, Jin-Hwan Yoon, Min-Hwa Suk, Yun-A Shin
ABSTRACT
The purpose of this study was to investigate the expression of lipogenesis- and lipolysis-related genes and proteins in skeletal muscles after 12 weeks of resistance training. Sprague-Dawley rats (n=12) were randomly divided into control (resting) and resistance training groups. A tower-climbing exercise, in which rats climbed to the top of their cage with a weight applied to their tails, used for resistance training. After 12 weeks, rats from the resistance training group had lower body weights (411.66±14.71 g vs. 478.33±24.63 g in the control), there was no significant difference between the two groups in the concentrations of total cholesterol, and high or low density lipoprotein cholesterol. However, the concentration of triglyceride was lower in resistance-trained rats (59.83±14.05 μg/mL vs 93.33±33.89 μg/mL in the control). The mRNA expression levels of the lipogenesis-related genes sterol regulatory element binding protein-1c, acetyl-CoA carboxylase, and fatty acid synthase were not significantly different between the resistance-trained and control rats; however, mRNA expression of the lipolysis-related carnitine palmitoyl transferase 1 and malonyl-CoA decarboxylase increased significantly with resistance training. AMP-activated protein kinase protein levels also significantly increased in resistance training group compared with in the control group. These results suggested that resistance exercise training contributing to reduced weight gain may be in part be due to increase the lipolysis metabolism and energy expenditure in response to resistance training.