- Effect of Lifestyle Intervention in the Concentration of Adipoquines and Branched Chain Amino Acids in Subjects with High Risk of Developing Type 2 Diabetes: Feel4Diabetes Study.
Effect of Lifestyle Intervention in the Concentration of Adipoquines and Branched Chain Amino Acids in Subjects with High Risk of Developing Type 2 Diabetes: Feel4Diabetes Study.
The global prevalence of type 2 diabetes (T2D) is increasing rapidly, especially in low- and middle-income countries and has a high number of associated comorbidities. Plasmatic concentrations of branched chain amino acids (BCAA) and retinol-binding protein 4 (RBP4) have been shown to be elevated in T2D subjects in cross-sectional studies. However, the effect of lifestyle community-based interventions on BCAA and RBP4 concentrations has not yet been analyzed. The Feel4Diabetes study is a school and community-based intervention that identified 360 European families with a high risk of developing T2D according to the FINDRISC questionnaire. Families were randomized in control and intervention groups were followed-up from 2016 to 2018. In the Spanish families, the concentration of BCAA and RBP4 was determined in 266 subjects (115 control and 151 intervention group) that attended the three time-point assessments by colorimetric and ELISA reaction, respectively. Baseline BCAA levels showed positive correlations with the FINDRISC score and glucose impairment (baseline glucose, insulin, and glycated hemoglobin), body mass index, and body weight. The participants receiving the community-based intervention showed a significant decrease in glycated hemoglobin and BCAA levels compared to the control group (p = 0.011 and p < 0.001, respectively). However, baseline RBP4 did not show significant correlations with anthropometric and glycemic parameters, and no significant change was observed in anthropometric parameters and RBP4 concentrations throughout the follow-up. A community-based intervention on lifestyle led to a significant reduction in BCAA levels regardless of weight loss. These findings suggest that this interventional approach could be promising in T2D prevention.