Merck
  • CD146 deficiency promotes plaque formation in a mouse model of atherosclerosis by enhancing RANTES secretion and leukocyte recruitment.
CD146 deficiency promotes plaque formation in a mouse model of atherosclerosis by enhancing RANTES secretion and leukocyte recruitment.
Journal of molecular and cellular cardiology (2019-04-01)
Muriel G Blin, Richard Bachelier, Karim Fallague, Karima Moussouni, Michel Aurrand-Lions, Samantha Fernandez, Benjamin Guillet, Stéphane Robert, Alexandrine Foucault-Bertaud, Nathalie Bardin, Marcel Blot-Chabaud, Françoise Dignat-George, Aurélie S Leroyer
ABSTRACT
The progression of atherosclerosis is based on the continued recruitment of leukocytes in the vessel wall. The previously described role of CD146 in leukocyte infiltration suggests an involvement for this adhesion molecule in the inflammatory response. In this study, we investigated the role of CD146 in leukocyte recruitment by using an experimental model of atherogenesis. The role of CD146 was explored in atherosclerosis by crossing CD146-/- mice with ApoE-/- mice. CD146 -/-/ApoE -/- and ApoE -/- mice were fed a Western diet for 24 weeks and were monitored for aortic wall thickness using high frequency ultrasound. The arterial wall was significantly thicker in CD146-deficient mice. After 24 weeks of Western diet, a significant increase of atheroma in both total aortic lesion and aortic sinus of CD146-null mice was observed. In addition, atherosclerotic lesions were more inflammatory since plaques from CD146-deficient mice contained more neutrophils and macrophages. This was due to up-regulation of RANTES secretion by macrophages in CD146-deficient atherosclerotic arteries. This prompted us to further address the function of CD146 in leukocyte recruitment during acute inflammation by using a second experimental model of peritonitis induced by thioglycollate. Neutrophil recruitment was significantly increased in CD146-deficient mice 12 h after peritonitis induction and associated with higher RANTES levels in the peritoneal cavity. In CD146-null macrophages, we also showed that increased RANTES production was dependent on constitutive inhibition of the p38-MAPK signaling pathway. Finally, Maraviroc, a RANTES receptor antagonist, was able to reduce atherosclerotic lesions and neutrophilia in CD146-deficient mice to the same level as that found in ApoE -/- mice. Our data indicate that CD146 deficiency is associated with the upregulation of RANTES production and increased inflammation of atheroma, which could influence the atherosclerotic plaque fate. Thus, these data identify CD146 agonists as potential new therapeutic candidates for atherosclerosis treatment.