Merck
  • Essential role of IkappaB kinase alpha in thymic organogenesis required for the establishment of self-tolerance.
Essential role of IkappaB kinase alpha in thymic organogenesis required for the establishment of self-tolerance.
Journal of immunology (Baltimore, Md. : 1950) (2006-03-21)
Dan Kinoshita, Fumiko Hirota, Tsuneyasu Kaisho, Michiyuki Kasai, Keisuke Izumi, Yoshimi Bando, Yasuhiro Mouri, Akemi Matsushima, Shino Niki, Hongwei Han, Kiyotaka Oshikawa, Noriyuki Kuroda, Masahiko Maegawa, Minoru Irahara, Kiyoshi Takeda, Shizuo Akira, Mitsuru Matsumoto
ABSTRACT
IkappaB kinase (IKK) alpha exhibits diverse biological activities through protein kinase-dependent and -independent functions, the former mediated predominantly through a noncanonical NF-kappaB activation pathway. The in vivo function of IKKalpha, however, still remains elusive. Because a natural strain of mice with mutant NF-kappaB-inducing kinase (NIK) manifests autoimmunity as a result of disorganized thymic structure with abnormal expression of Rel proteins in the thymic stroma, we speculated that the NIK-IKKalpha axis might constitute an essential step in the thymic organogenesis that is required for the establishment of self-tolerance. An autoimmune disease phenotype was induced in athymic nude mice by grafting embryonic thymus from IKKalpha-deficient mice. The thymic microenvironment that caused autoimmunity in an IKKalpha-dependent manner was associated with defective processing of NF-kappaB2, resulting in the impaired development of thymic epithelial cells. Thus, our results demonstrate a novel function for IKKalpha in thymic organogenesis for the establishment of central tolerance that depends on its protein kinase activity in cooperation with NIK.