Merck
  • Enzyme immobilization on glass fiber membrane for detection of halogenated compounds.
Enzyme immobilization on glass fiber membrane for detection of halogenated compounds.
Analytical biochemistry (2020-09-27)
Ijaz Gul, Qian Wang, Qifa Jiang, Ruiqin Fang, Lixia Tang
ABSTRACT
Enzyme immobilization using inorganic membranes has enticed increased attention as they not only improve enzyme stability, but also furnish user-friendly biodevices that can be tailored to different applications. Herein, we explored the suitability of the glass fiber membrane for enzyme immobilization and its application for halocarbon detection. For this, halohydrin dehalogenase (HheC) and bovine serum albumin were crosslinked and immobilized on a glass fiber membrane without membrane functionalization. Immobilized HheC exhibited higher storage stability than its free counterpart over 60 days at 4 °C (67% immobilized vs. 8.1% free) and 30 °C (77% immobilized vs. 57% free). Similarly, the thermal endurance of the immobilized HheC was significantly improved. The practical utility of the membrane-immobilized enzyme was demonstrated by colorimetric detection of 1,3-dichloro-2-propanol (1,3-DCP) and 2,3-dibromo-1-propanol (2,3-DBP) as model analytes. Under optimized conditions, the detection limits of 0.06 mM and 0.09 mM were achieved for 1,3-DCP and 2,3-DBP, respectively. The satisfactory recoveries were observed with spiked river and lake water samples, which demonstrate the application potential of immobilized HheC for screening contaminants in water samples. Our results revealed that the proposed frugal and facile approach could be useful for enzyme stabilization, and mitigation of halocarbon pollution.
Sigma-Aldrich
2,3-Dibromo-1-propanol, 98%