Merck
  • Asbestos induces nitric oxide synthesis in mesothelioma cells via Rho signaling inhibition.
Asbestos induces nitric oxide synthesis in mesothelioma cells via Rho signaling inhibition.
American journal of respiratory cell and molecular biology (2007-02-27)
Chiara Riganti, Sara Orecchia, Francesca Silvagno, Gianpiero Pescarmona, Pier Giacomo Betta, Elena Gazzano, Elisabetta Aldieri, Dario Ghigo, Amalia Bosia
ABSTRACT
We have observed that in three human malignant mesothelioma cell lines, crocidolite asbestos induced the activation of the transcription factor NF-kappaB and the synthesis of nitric oxide (NO) by inhibiting the RhoA signaling pathway. The incubation with crocidolite decreased the level of GTP-bound RhoA and the activity of Rho-dependent kinase, and induced the activation of Akt/PKB and IkBalpha kinase, leading to the nuclear translocation of NF-kappaB. The effects of crocidolite fibers on NF-kappaB activation and NO synthesis were mimicked by Y27632 (an inhibitor of the Rho-dependent kinases) and toxin B (an inhibitor of RhoA GTPase activity), while they were reverted by mevalonic acid, the product of 3-hydroxy-3-methylglutaryl coenzyme A (HMGCoA) reductase. Furthermore, crocidolite, similarly to mevastatin, inhibited the synthesis of cholesterol and ubiquinone and the prenylation of RhoA: these effects were prevented in the presence of mevalonic acid. This suggests that crocidolite fibers might inhibit the synthesis of isoprenoid molecules at the level of the HMGCoA reductase reaction or of an upstream step, thus impairing the prenylation and subsequent activation of RhoA. Akt can stimulate NO synthesis via a double mechanism: it can activate the inducible NO synthase via the NF-kappaB pathway and the endothelial NO synthase via a direct phosphorylation. Our results suggest that crocidolite increases the NO levels in mesothelioma cells by modulating both NO synthase isoforms.