Merck
  • Characterization of a human monoclonal antibody generated from a B-cell specific for a prefusion-stabilized spike protein of Middle East respiratory syndrome coronavirus.
Characterization of a human monoclonal antibody generated from a B-cell specific for a prefusion-stabilized spike protein of Middle East respiratory syndrome coronavirus.
PloS one (2020-05-10)
Jang-Hoon Choi, Hye-Min Woo, Tae-Young Lee, So-Young Lee, Sang-Mu Shim, Woo-Jung Park, Jeong-Sun Yang, Joo Ae Kim, Mi-Ran Yun, Dae-Won Kim, Sung Soon Kim, Yi Zhang, Wei Shi, Lingshu Wang, Barney S Graham, John R Mascola, Nanshuang Wang, Jason S McLellan, Joo-Yeon Lee, Hansaem Lee
ABSTRACT
Middle East respiratory syndrome coronavirus (MERS-CoV) causes severe respiratory infection and continues to infect humans, thereby contributing to a high mortality rate (34.3% in 2019). In the absence of an available licensed vaccine and antiviral agent, therapeutic human antibodies have been suggested as candidates for treatment. In this study, human monoclonal antibodies were isolated by sorting B cells from patient's PBMC cells with prefusion stabilized spike (S) probes and a direct immunoglobulin cloning strategy. We identified six receptor-binding domain (RBD)-specific and five S1 (non-RBD)-specific antibodies, among which, only the RBD-specific antibodies showed high neutralizing potency (IC50 0.006-1.787 μg/ml) as well as high affinity to RBD. Notably, passive immunization using a highly potent antibody (KNIH90-F1) at a relatively low dose (2 mg/kg) completely protected transgenic mice expressing human DPP4 against MERS-CoV lethal challenge. These results suggested that human monoclonal antibodies isolated by using the rationally designed prefusion MERS-CoV S probe could be considered potential candidates for the development of therapeutic and/or prophylactic antiviral agents for MERS-CoV human infection.