Merck
  • 5-HT1A targeting PARCEST agent DO3AM-MPP with potential for receptor imaging: Synthesis, physico-chemical and MR studies.
5-HT1A targeting PARCEST agent DO3AM-MPP with potential for receptor imaging: Synthesis, physico-chemical and MR studies.
Bioorganic chemistry (2020-12-20)
Anju, Shubhra Chaturvedi, Vishakha Chaudhary, Pradeep Pant, Preeti Jha, Senthil S Kumaran, Firasat Hussain, Anil Kumar Mishra
ABSTRACT
Contrast enhancement in MRI using magnetization or saturation transfer techniques promises better sensitivity, and faster acquisition compared to T1 or T2 contrast. This work reports the synthesis and evaluation of 5-HT1A targeted PARACEST MRI contrast agent using 1,4,7,10-tetraazacycloDOdecane-4,7,10-triacetAMide (DO3AM) as the bifunctional chelator, and 5-HT1A-antagonist methoxyphenyl piperazine (MPP) as a targeting unit. The multi-step synthesis led to the MPP conjugated DO3AM with 60% yield. CEST-related physicochemical parameters were evaluated after loading DO3AM-MPP with paramagnetic MRI active lanthanides: Gadolinium (Gd-DO3AM-MPP) and Europium (Eu-DO3AM-MPP). Luminescence lifetime measurements with Eu-DO3AM-MPP and computational DFT studies using Gd-DO3AM-MPP revealed the coordination of one water molecule (q = 1.43) with metal-water distance (rM-H2O) of 2.7 Å and water residence time (τm) of 0.23 ms. The dissociation constant of Kd 62 ± 0.02 pM as evaluated from fluorescence quenching of 5-HT1A (protein) and docking score of -4.81 in theoretical evaluation reflect the binding potential of the complex Gd-DO3AM-MPP with the receptor 5-HT1A. Insights of the docked pose reflect the importance of NH2 (amide) and aromatic ring in Gd-DO3AM-MPP while interacting with Ser 374 and Phe 370 in the antagonist binding pocket of 5-HT1A. Gd-DO3AM-MPP shows longitudinal relaxivity 5.85 mM-1s-1 with a water residence lifetime of 0.93 ms in hippocampal homogenate containing 5-HT1A. The potentiometric titration of DO3AM-MPP showed strong selectivity for Gd3+ over physiological metal ions such as Zn2+ and Cu2+. The in vitro and in vivo studies confirmed the minimal cytotoxicity and presential binding of Gd-DO3AM-MPP with 5-HT1A receptor in the hippocampus region of the mice. Summarizing, the complex Gd-DO3AM-MPP can have a potential for CEST imaging of 5-HT1A receptors.
Sigma-Aldrich
Cyclen, 97%