Merck
  • A microbubble-sparged yeast propagation-fermentation process for bioethanol production.
A microbubble-sparged yeast propagation-fermentation process for bioethanol production.
Biotechnology for biofuels (2020-06-12)
Vijayendran Raghavendran, Joseph P Webb, Michaël L Cartron, Vicki Springthorpe, Tony R Larson, Michael Hines, Hamza Mohammed, William B Zimmerman, Robert K Poole, Jeffrey Green
ABSTRACT
Industrial biotechnology will play an increasing role in creating a more sustainable global economy. For conventional aerobic bioprocesses supplying O2 can account for 15% of total production costs. Microbubbles (MBs) are micron-sized bubbles that are widely used in industry and medical imaging. Using a fluidic oscillator to generate energy-efficient MBs has the potential to decrease the costs associated with aeration. However, little is understood about the effect of MBs on microbial physiology. To address this gap, a laboratory-scale MB-based Saccharomyces cerevisiae Ethanol Red propagation-fermentation bioethanol process was developed and analysed. Aeration with MBs increased O2 transfer to the propagation cultures. Titres and yields of bioethanol in subsequent anaerobic fermentations were comparable for MB-propagated and conventional, regular bubble (RB)-propagated yeast. However, transcript profiling showed significant changes in gene expression in the MB-propagated yeast compared to those propagated using RB. These changes included up-regulation of genes required for ergosterol biosynthesis. Ergosterol contributes to ethanol tolerance, and so the performance of MB-propagated yeast in fed-batch fermentations sparged with 1% O2 as either RBs or MBs were tested. The MB-sparged yeast retained higher levels of ergosteryl esters during the fermentation phase, but this did not result in enhanced viability or ethanol production compared to ungassed or RB-sparged fermentations. The performance of yeast propagated using energy-efficient MB technology in bioethanol fermentations is comparable to that of those propagated conventionally. This should underpin the future development of MB-based commercial yeast propagation.