Merck
  • Reduction of Salt and Fat in Frankfurter Sausages by Addition of Agaricus bisporus and Pleurotus ostreatus Flour.
Reduction of Salt and Fat in Frankfurter Sausages by Addition of Agaricus bisporus and Pleurotus ostreatus Flour.
Foods (Basel, Switzerland) (2020-06-13)
Magdalena I Cerón-Guevara, Esmeralda Rangel-Vargas, José M Lorenzo, Roberto Bermúdez, Mirian Pateiro, Jose A Rodríguez, Irais Sánchez-Ortega, Eva M Santos
ABSTRACT
The reduction of fat and salt and the incorporation of fiber-rich compounds in frankfurters is a trend to improve their nutritional profile. The objective of this study was to evaluate the partial replacement of 30 and 50% of pork backfat and 50% of salt by adding edible mushroom flour (2.5 and 5%) from Agaricus bisporus (Ab) and Pleurotus ostreatus (Po) on physicochemical, microbiological and sensory properties of frankfurters sausages during cold storage. The addition of flours increased the moisture, and the dietary fiber contents in frankfurters, keeping the amino acid profile. Lipid oxidation remained under acceptable values despite not antioxidant effect was observed by mushrooms flours. Only spore-forming bacteria were found during cold storage. Color and texture was modified by addition of mushroom, being the Ab samples darker, while Po flour addition resulted in softer and less cohesive sausages. Although lower color, flavor, and taste scores were given to the mushroom samples than the control, they ranked in the acceptable level confirming that the inclusion of 2.5 and 5% of Ab and Po flours in fat- and salt-reduced frankfurter sausages resulted a feasible strategy to enhance the nutritional profile these products.
Product Description
Supelco
Total Dietary Fiber Assay Kit, sufficient for ~100 assays