Merck
  • Clinical Characteristics and Treatment Outcomes of Pathologically Confirmed Aspergillus Nodules.
Clinical Characteristics and Treatment Outcomes of Pathologically Confirmed Aspergillus Nodules.
Journal of clinical medicine (2020-07-16)
Noeul Kang, Jiyeon Park, Byung Woo Jhun
ABSTRACT
Aspergillus nodules represent a subtype of chronic pulmonary aspergillosis, but details on their characteristics and outcomes are limited. We evaluated 80 patients with pathologically confirmed Aspergillus nodules between January 2009 and December 2016. The median age of the patients was 59 years, and 46 (58%) were women. Seventy-three (91%) patients were surgically diagnosed with Aspergillus nodules and the remaining seven (9%) patients were diagnosed by percutaneous transthoracic needle biopsy. The median long-axis diameter of nodules was 22 mm, and nodules had an internal cavity in 49 (61%) patients. Spiculation and calcification were observed in 20% and 39% of patients, respectively. Ninety percent (18/20) of nodules showed uptake on positron emission tomography. Serum Aspergillus precipitin IgG antibody was positive in 42% (10/24) of tested patients. Seventy-three (91%) patients underwent surgery without (n = 58) or with (n = 15) adjuvant antifungal therapy, and the remaining seven (9%) patients received antifungal therapy alone (n = 5) or no treatment (n = 2). Three patients experienced postoperative pulmonary complications: pneumothorax, hemoptysis, and acute lung injury (n = 1 each). There was no recurrence during the median follow-up period of 36.8 months. In conclusion, surgery could be a treatment strategy worth considering for most Aspergillus nodules. However, given that our study population was heterogeneous, further well-designed studies are need.
Sigma-Aldrich
Human IgG ELISA Kit, for serum, plasma