Merck
  • α-Tricalcium phosphate: synthesis, properties and biomedical applications.
α-Tricalcium phosphate: synthesis, properties and biomedical applications.
Acta biomaterialia (2011-06-30)
R G Carrodeguas, S De Aza
ABSTRACT
Nowadays, α-tricalcium phosphate (α-TCP, α-Ca(3)(PO(4))(2)) is receiving growing attention as a raw material for several injectable hydraulic bone cements, biodegradable bioceramics and composites for bone repair. In the phase equilibrium diagram of the CaO-P(2)O(5) system, three polymorphs corresponding to the composition Ca(3)(PO(4))(2) are recognized: β-TCP, α-TCP and α'-TCP. α-TCP is formed by heating the low-temperature polymorph β-TCP or by thermal crystallization of amorphous precursors with the proper composition above the transformation temperature. The α-TCP phase may be retained at room temperature in a metastable state, and its range of stability is strongly influenced by ionic substitutions. It is as biocompatible as β-TCP, but more soluble, and hydrolyses rapidly to calcium-deficient hydroxyapatite, which makes α-TCP a useful component for preparing self-setting osteotransductive bone cements and biodegradable bioceramics and composites for bone repairing. The literature published on the synthesis and properties of α-TCP is sometimes contradictory, and therefore this article focuses on reviewing and critically discussing the synthetic methods and physicochemical and biological properties of α-TCP-based biomaterials (excluding α-TCP-based bone cements).
Sigma-Aldrich
Alpha-Tricalcium phosphate, (TCP), powder, ≥98%, <500 μm particle size
Pricing and availability is not currently available.