Merck
  • Quinoline Based Monocarbonyl Curcumin Analogs as Potential Antifungal and Antioxidant Agents: Synthesis, Bioevaluation and Molecular Docking Study.
Quinoline Based Monocarbonyl Curcumin Analogs as Potential Antifungal and Antioxidant Agents: Synthesis, Bioevaluation and Molecular Docking Study.
Chemistry & biodiversity (2019-12-22)
Amol A Nagargoje, Satish V Akolkar, Madiha M Siddiqui, Dnyaneshwar D Subhedar, Jaiprakash N Sangshetti, Vijay M Khedkar, Bapurao B Shingate
ABSTRACT
In search for new fungicidal and free radical scavenging agents, we synthesized a focused library of 2-chloroquinoline based monocarbonyl analogs of curcumin (MACs). The synthesized MACs were evaluated for in vitro antifungal and antioxidant activity. The antifungal activity was evaluated against five different fungal strains such as Candida albicans, Fusarium oxysporum, Aspergillus flavus, Aspergillus niger, and Cryptococcus neoformans, respectively. Most of the synthesized MACs displayed promising antifungal activity compared to the standard drug Miconazole. Furthermore, molecular docking study on a crucial fungal enzyme sterol 14α-demethylase (CYP51) could provide insight into the plausible mechanism of antifungal activity. MACs were also screened for in vitro radical scavenging activity using butylated hydroxytoluene (BHT) as a standard. Almost all MACs exhibited better antioxidant activity compared to BHT.
Sigma-Aldrich
2-Chloroquinoline, 99%