Merck
  • Expression, purification, and characterization of human mannose-6-phosphate receptor - Extra cellular domain from a stable cell line utilizing a small molecule biomimetic of the mannose-6-phosphate moiety.
Expression, purification, and characterization of human mannose-6-phosphate receptor - Extra cellular domain from a stable cell line utilizing a small molecule biomimetic of the mannose-6-phosphate moiety.
Protein expression and purification (2020-02-07)
Brian Dwyer, Dianna Lundberg, Andrea Iskenderian, Bettina Strack-Logue, Brian Pescatore, Angela W Norton, Jin Xu, Muthuraman Meiyappan, Michael F Concino, Bohong Zhang
ABSTRACT
The cation-independent mannose-6-phosphate receptor (CI-M6PR, aka insulin-like growth factor II receptor or IGFIIR) is a membrane protein that plays a central role in the trafficking of lysosomal acid hydrolases into lysosomes via mannose-6-phosphate (M6P) binding domains. In order to maintain cellular metabolic/catabolic homeostasis, newly synthesized lysosomal acid hydrolases are required to bind to M6PR for transit. Acid hydrolases secreted by cells can also be internalized via M6PR residing on the cell membrane and are transported to the lysosomes, a feature that enables enzyme replacement therapy for the treatment of several lysosomal storage disorders. Therefore, a thorough characterization of this receptor is critical to the development of lysosomal enzyme-based therapeutics that utilize M6PR for drug delivery to the lysosome. However, the extracellular domain (ECD) of M6PR is highly complex, containing 15-mannose receptor homology (MRH) domains. In addition, homodimerization of the receptor can occur at the membrane, making its characterization challenging. In this study, a novel human M6PR (hM6PR)-overexpressing cell line originally established for hM6PR cellular uptake assay was utilized for production of hM6PR-ECD, and a novel small molecule biomimetic (aminophenyl-M6P) affinity resin was developed for the purification of M6PR-ECD. The affinity-purified hM6PR-ECD was monomeric, contained 14 intact MRH domains (1-14) and a partial MRH domain 15, and was successfully employed in ELISA-based and surface plasmon resonance-based binding assays to demonstrate its ligand-binding functionality, making it suitable for the evaluation of biotherapeutics that utilize M6PR for cellular internalization.
Sigma-Aldrich
Mouse IGF-II ELISA Kit, for cell culture supernatant