- Analysis of potential response predictors to capecitabine/temozolomide in metastatic pancreatic neuroendocrine tumors.
Analysis of potential response predictors to capecitabine/temozolomide in metastatic pancreatic neuroendocrine tumors.
The capecitabine and temozolomide (CAPTEM) regimen is active in the treatment of metastatic pancreatic neuroendocrine tumors (pNETs), with response rates ranging from 30 to 70%. Small retrospective studies suggest that O(6)-methylguanine DNA methyltransferase (MGMT) deficiency predicts response to temozolomide. High tumor proliferative activity is also commonly perceived as a significant predictor of response to cytotoxic chemotherapy. It is unclear whether chromosomal instability (CIN), which correlates with alternative lengthening of telomeres (ALT), is a predictive factor. In this study, we evaluated 143 patients with advanced pNET who underwent treatment with CAPTEM for radiographic and biochemical response. MGMT expression (n=52), grade (n=128) and ALT activation (n=46) were investigated as potential predictive biomarkers. Treatment with CAPTEM was associated with an overall response rate (ORR) of 54% by RECIST 1.1. Response to CAPTEM was not influenced by MGMT expression, proliferative activity or ALT pathway activation. Based on these results, no biomarker-driven selection criteria for use of the CAPTEM regimen can be recommended at this time.