Merck
  • Inhibition of PMCA activity by tau as a function of aging and Alzheimer's neuropathology.
Inhibition of PMCA activity by tau as a function of aging and Alzheimer's neuropathology.
Biochimica et biophysica acta (2015-04-22)
María Berrocal, Isaac Corbacho, María Vázquez-Hernández, Jesús Ávila, M Rosario Sepúlveda, Ana M Mata
ABSTRACT
Ca2+-ATPases are plasma membrane and intracellular membrane transporters that use the energy of ATP hydrolysis to pump cytosolic Ca2+ out of the cell (PMCA) or into internal stores. These pumps are the main high-affinity Ca2+ systems involved in the maintenance of intracellular free Ca2+ at the properly low level in eukaryotic cells. The failure of neurons to keep optimal intracellular Ca2+ concentrations is a common feature of neurodegeneration by aging and aging-linked neuropathologies, such as Alzheimer's disease (AD). This disease is characterized by the accumulation of β-amyloid senile plaques and neurofibrillary tangles of tau, a protein that plays a key role in axonal transport. Here we show a novel inhibition of PMCA activity by tau which is concentration-dependent. The extent of inhibition significantly decreases with aging in mice and control human brain membranes, but inhibition profiles were similar in AD-affected brain membrane preparations, independently of age. No significant changes in PMCA expression and localization with aging or neuropathology were found. These results point out a link between Ca2+-transporters, aging and neurodegeneration mediated by tau protein.