Merck
  • The PYRIN domain-only protein POP3 inhibits ALR inflammasomes and regulates responses to infection with DNA viruses.
The PYRIN domain-only protein POP3 inhibits ALR inflammasomes and regulates responses to infection with DNA viruses.
Nature immunology (2014-02-18)
Sonal Khare, Rojo A Ratsimandresy, Lúcia de Almeida, Carla M Cuda, Stephanie L Rellick, Alexander V Misharin, Melissa C Wallin, Anu Gangopadhyay, Eleonora Forte, Eva Gottwein, Harris Perlman, John C Reed, David R Greaves, Andrea Dorfleutner, Christian Stehlik
ABSTRACT
The innate immune system responds to infection and tissue damage by activating cytosolic sensory complexes called 'inflammasomes'. Cytosolic DNA is sensed by AIM2-like receptors (ALRs) during bacterial and viral infections and in autoimmune diseases. Subsequently, recruitment of the inflammasome adaptor ASC links ALRs to the activation of caspase-1. A controlled immune response is crucial for maintaining homeostasis, but the regulation of ALR inflammasomes is poorly understood. Here we identified the PYRIN domain (PYD)-only protein POP3, which competes with ASC for recruitment to ALRs, as an inhibitor of DNA virus-induced activation of ALR inflammasomes in vivo. Data obtained with a mouse model with macrophage-specific POP3 expression emphasize the importance of the regulation of ALR inflammasomes in monocytes and macrophages.