Merck
  • Salmonella infection induces recruitment of Caspase-8 to the inflammasome to modulate IL-1β production.
Salmonella infection induces recruitment of Caspase-8 to the inflammasome to modulate IL-1β production.
Journal of immunology (Baltimore, Md. : 1950) (2013-10-15)
Si Ming Man, Panagiotis Tourlomousis, Lee Hopkins, Tom P Monie, Katherine A Fitzgerald, Clare E Bryant
ABSTRACT
Nucleotide-binding oligomerization domain-like receptors (NLRs) detect pathogens and danger-associated signals within the cell. Salmonella enterica serovar Typhimurium, an intracellular pathogen, activates caspase-1 required for the processing of the proinflammatory cytokines, pro-IL-1β and pro-IL-18, and pyroptosis. In this study, we show that Salmonella infection induces the formation of an apoptosis-associated specklike protein containing a CARD (ASC)-Caspase-8-Caspase-1 inflammasome in macrophages. Caspase-8 and caspase-1 are recruited to the ASC focus independently of one other. Salmonella infection initiates caspase-8 proteolysis in a manner dependent on NLRC4 and ASC, but not NLRP3, caspase-1 or caspase-11. Caspase-8 primarily mediates the synthesis of pro-IL-1β, but is dispensable for Salmonella-induced cell death. Overall, our findings highlight that the ASC inflammasome can recruit different members of the caspase family to induce distinct effector functions in response to Salmonella infection.