Merck
  • Impaired ABCA1/ABCG1-mediated lipid efflux in the mouse retinal pigment epithelium (RPE) leads to retinal degeneration.
Impaired ABCA1/ABCG1-mediated lipid efflux in the mouse retinal pigment epithelium (RPE) leads to retinal degeneration.
eLife (2019-03-14)
Federica Storti, Katrin Klee, Vyara Todorova, Regula Steiner, Alaa Othman, Saskia van der Velde-Visser, Marijana Samardzija, Isabelle Meneau, Maya Barben, Duygu Karademir, Valda Pauzuolyte, Sanford L Boye, Frank Blaser, Christoph Ullmer, Joshua L Dunaief, Thorsten Hornemann, Lucia Rohrer, Anneke den Hollander, Arnold von Eckardstein, Jürgen Fingerle, Cyrille Maugeais, Christian Grimm
ABSTRACT
Age-related macular degeneration (AMD) is a progressive disease of the retinal pigment epithelium (RPE) and the retina leading to loss of central vision. Polymorphisms in genes involved in lipid metabolism, including the ATP-binding cassette transporter A1 (ABCA1), have been associated with AMD risk. However, the significance of retinal lipid handling for AMD pathogenesis remains elusive. Here, we study the contribution of lipid efflux in the RPE by generating a mouse model lacking ABCA1 and its partner ABCG1 specifically in this layer. Mutant mice show lipid accumulation in the RPE, reduced RPE and retinal function, retinal inflammation and RPE/photoreceptor degeneration. Data from human cell lines indicate that the ABCA1 AMD risk-conferring allele decreases ABCA1 expression, identifying the potential molecular cause that underlies the genetic risk for AMD. Our results highlight the essential homeostatic role for lipid efflux in the RPE and suggest a pathogenic contribution of reduced ABCA1 function to AMD.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid