Merck
  • Polymer-Mediated Inhibition of Pro-invasive Nucleic Acid DAMPs and Microvesicles Limits Pancreatic Cancer Metastasis.
Polymer-Mediated Inhibition of Pro-invasive Nucleic Acid DAMPs and Microvesicles Limits Pancreatic Cancer Metastasis.
Molecular therapy : the journal of the American Society of Gene Therapy (2018-03-20)
Ibtehaj Naqvi, Ruwan Gunaratne, Jessica E McDade, Angelo Moreno, Rachel E Rempel, Douglas C Rouse, Silvia Gabriela Herrera, David S Pisetsky, Jaewoo Lee, Rebekah R White, Bruce A Sullenger
ABSTRACT
Nucleic acid binding polymers (NABPs) have been extensively used as vehicles for DNA and RNA delivery. More recently, we discovered that a subset of these NABPs can also serve as anti-inflammatory agents by capturing pro-inflammatory extracellular nucleic acids and associated protein complexes that promote activation of toll-like receptors (TLRs) in diseases such as lupus erythematosus. Nucleic-acid-mediated TLR signaling also facilitates tumor progression and metastasis in several cancers, including pancreatic cancer (PC). In addition, extracellular DNA and RNA circulate on or within lipid microvesicles, such as microparticles or exosomes, which also promote metastasis by inducing pro-tumorigenic signaling in cancer cells and pre-conditioning secondary sites for metastatic establishment. Here, we explore the use of an NABP, the 3rd generation polyamidoamine dendrimer (PAMAM-G3), as an anti-metastatic agent. We show that PAMAM-G3 not only inhibits nucleic-acid-mediated activation of TLRs and invasion of PC tumor cells in vitro, but can also directly bind extracellular microvesicles to neutralize their pro-invasive effects as well. Moreover, we demonstrate that PAMAM-G3 dramatically reduces liver metastases in a syngeneic murine model of PC. Our findings identify a promising therapeutic application of NABPs for combating metastatic disease in PC and potentially other malignancies.