Merck
  • The orientation and dynamics of estradiol and estradiol oleate in lipid membranes and HDL disc models.
The orientation and dynamics of estradiol and estradiol oleate in lipid membranes and HDL disc models.
Biophysical journal (2014-07-06)
Alexander Vogel, Holger A Scheidt, Scott E Feller, Jari Metso, Robert M Badeau, Matti J Tikkanen, Kristiina Wähälä, Matti Jauhiainen, Daniel Huster
ABSTRACT
Estradiol (E2) and E2 oleate associate with high-density lipoproteins (HDLs). Their orientation in HDLs is unknown. We studied the orientation of E2 and E2 oleate in membranes and reconstituted HDLs, finding that E2 and E2 oleate are membrane-associated and highly mobile. Our combination of NMR measurements, molecular dynamics simulation, and analytic theory identifies three major conformations where the long axis of E2 assumes a parallel, perpendicular, or antiparallel orientation relative to the membrane's z-direction. The perpendicular orientation is preferred, and furthermore, in this orientation, E2 strongly favors a particular roll angle, facing the membrane with carbons 6, 7, 15, and 16, whereas carbons 1, 2, 11, and 12 point toward the aqueous phase. In contrast, the long axis of E2 oleate is almost exclusively oriented at an angle of ∼60° to the z-direction. In such an orientation, the oleoyl chain is firmly inserted into the membrane. Thus, both E2 and E2 oleate have a preference for interface localization in the membrane. These orientations were also found in HDL discs, suggesting that only lipid-E2 interactions determine the localization of the molecule. The structural mapping of E2 and E2 oleate may provide a design platform for specific E2-HDL-targeted pharmacological therapies.
Sigma-Aldrich
L-α-Phosphatidylcholine, egg yolk, Type XI-E, 100 mg/mL in chloroform, ≥99%, solution