Merck
  • Berberine alleviates insulin resistance by reducing peripheral branched-chain amino acids.
Berberine alleviates insulin resistance by reducing peripheral branched-chain amino acids.
American journal of physiology. Endocrinology and metabolism (2018-11-14)
Shi-Jun Yue, Juan Liu, Ai-Ting Wang, Xin-Tong Meng, Zhi-Rui Yang, Cheng Peng, Hua-Shi Guan, Chang-Yun Wang, Dan Yan
ABSTRACT
Increased circulating branched-chain amino acids (BCAAs) have been involved in the pathogenesis of obesity and insulin resistance (IR). However, evidence relating berberine (BBR), gut microbiota, BCAAs, and IR is limited. Here, we showed that BBR could effectively rectify steatohepatitis and glucose intolerance in high-fat diet (HFD)-fed mice. BBR reorganized gut microbiota populations under both the normal chow diet (NCD) and HFD. Particularly, BBR noticeably decreased the relative abundance of BCAA-producing bacteria, including order Clostridiales; families Streptococcaceae, Clostridiaceae, and Prevotellaceae; and genera Streptococcus and Prevotella. Compared with the HFD group, predictive metagenomics indicated a reduction in the proportion of gut microbiota genes involved in BCAA biosynthesis but the enrichment genes for BCAA degradation and transport by BBR treatment. Accordingly, the elevated serum BCAAs of HFD group were significantly decreased by BBR. Furthermore, the Western blotting results implied that BBR could promote the BCAA catabolism in the liver and epididymal white adipose tissues of HFD-fed mice by activation of the multienzyme branched-chain α-ketoacid dehydrogenase complex (BCKDC), whereas by inhibition of the phosphorylation state of BCKDHA (E1α subunit) and branched-chain α-ketoacid dehydrogenase kinase (BCKDK). The ex vivo assay further confirmed that BBR could increase BCAA catabolism in both AML12 hepatocytes and 3T3-L1 adipocytes. Finally, data from healthy subjects and diabetics confirmed that BBR could improve glycemic control and modulate circulating BCAAs. Together, our findings clarified BBR improving IR associated not only with gut microbiota alteration in BCAA biosynthesis but also with BCAA catabolism in liver and adipose tissues.
Sigma-Aldrich
Anti-DBT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2