Merck
  • Nickel@Siloxene catalytic nanosheets for high-performance CO2 methanation.
Nickel@Siloxene catalytic nanosheets for high-performance CO2 methanation.
Nature communications (2019-06-15)
Xiaoliang Yan, Wei Sun, Liming Fan, Paul N Duchesne, Wu Wang, Christian Kübel, Di Wang, Sai Govind Hari Kumar, Young Feng Li, Alexandra Tavasoli, Thomas E Wood, Darius L H Hung, Lili Wan, Lu Wang, Rui Song, Jiuli Guo, Ilya Gourevich, Abdinoor A Jelle, Jingjun Lu, Ruifeng Li, Benjamin D Hatton, Geoffrey A Ozin
ABSTRACT
Two-dimensional (2D) materials are of considerable interest for catalyzing the heterogeneous conversion of CO2 to synthetic fuels. In this regard, 2D siloxene nanosheets, have escaped thorough exploration, despite being composed of earth-abundant elements. Herein we demonstrate the remarkable catalytic activity, selectivity, and stability of a nickel@siloxene nanocomposite; it is found that this promising catalytic performance is highly sensitive to the location of the nickel component, being on either the interior or the exterior of adjacent siloxene nanosheets. Control over the location of nickel is achieved by employing the terminal groups of siloxene and varying the solvent used during its nucleation and growth, which ultimately determines the distinct reaction intermediates and pathways for the catalytic CO2 methanation. Significantly, a CO2 methanation rate of 100 mmol gNi-1 h-1 is achieved with over 90% selectivity when nickel resides specifically between the sheets of siloxene.