Merck
  • Left ventricular mass estimation by real-time 3D echocardiography favourably competes with CMR in congenital left ventricular disease.
Left ventricular mass estimation by real-time 3D echocardiography favourably competes with CMR in congenital left ventricular disease.
Scientific reports (2019-08-17)
Miriam Michel, Wael Shabanah, Hermann Körperich, Andrea Kelter-Klöpping, Andreas Entenmann, Anca Racolta, Kai Thorsten Laser
ABSTRACT
Assessment of left ventricular mass (LVM) is important in the evaluation of patients with congenital heart disease (CHD) and cardiac magnetic resonance imaging (CMR) is the gold standard. Recent software allows LVM calculation by real-time 3-dimensional echocardiography (RT3DE). We investigated the impact of different software analysis tools on LVM determination by CMR or RT3DE in a cohort of patients with heterogeneous left ventricular (LV) disease. 37 subjects (17 patients, mean age 18.7 y; 20 controls, mean age 13.2 y) underwent CMR and RT3DE. CMR LVM and RT3DE calculations were done using two different LV-analysis software packages for each modality: CMR i) customized software "CMR HDZ", CMR ii) "CMR ISP"; RT3DE i) "Toshiba", RT3DE ii) "Tomtec", 4D LV-Analysis Version 3.1 (built 3.1.0.258661). Intra- and interobserver variabilities were calculated. Only RT3DE-derived LVM showed significant software-dependent differences. RT3DE-derived LVM (both softwares) was significantly higher than CMR-derived LVM (both softwares). The two different methods and four evaluation software packages for LVM assessment were well correlated with each other. Intra- and interobserver variability of LVM as assessed by each single modality or software was low. Despite software dependency and overestimation of RT3DE-assessed LVM by 5 to 10%, RT3DE still competes with the gold standard, CMR, even in patients with various forms of LV disease. The use of optimized software, especially for RT3DE, should improve the accuracy of LVM assessment, overcoming LVM overestimation.