Merck
  • Enhancement of synaptic plasticity and reversal of impairments in motor and cognitive functions in a mouse model of Angelman Syndrome by a small neurogenic molecule, NSI-189.
Enhancement of synaptic plasticity and reversal of impairments in motor and cognitive functions in a mouse model of Angelman Syndrome by a small neurogenic molecule, NSI-189.
Neuropharmacology (2018-11-09)
Yan Liu, Karl Johe, Jiandong Sun, Xiaoning Hao, Yubin Wang, Xiaoning Bi, Michel Baudry
ABSTRACT
NSI-189 Phosphate, (4-benzylpiperazin-1-yl)-[2-(3-methyl-butylamino)pyridin-3-yl] methanone is a new chemical entity under development for the treatment of MDD, based upon preclinical data demonstrating stimulation of neurogenesis of human hippocampus-derived neural stem cells in vitro and in mouse hippocampus in vivo. Previous studies have examined the tolerability and efficacy of NSI-189 for treating major depressive disorder (MDD). NSI-189 has shown significant potential as a treatment for MDD, with concurrent improvement of a cognition scale in a small double-blind, placebo-controlled study. The current study evaluated its possible application for the treatment of Angelman Syndrome. Incubation of acute hippocampal slices from wild-type mice with NSI-189 resulted in a time- and dose-dependent increase in the magnitude of long-term potentiation (LTP) elicited by theta burst stimulation (TBS). The same protocol enhanced TBS-induced LTP in acute hippocampal slices from AS mice. A short treatment with daily injections of NSI-189 in AS mice reversed impairments in cognitive and motor functions, while it slightly enhanced performance of WT mice. The effects of NSI-189 on synaptic plasticity and cognitive functions were associated with activation of the TrkB and Akt pathways. These results suggest that NSI-189 could represent a potential treatment for AS patients.