Merck
  • Serine (threonine) phosphatase(s) acting on cAMP-dependent phosphoproteins in mammalian mitochondria.
Serine (threonine) phosphatase(s) acting on cAMP-dependent phosphoproteins in mammalian mitochondria.
FEBS letters (2002-02-20)
Anna Signorile, Anna Maria Sardanelli, Rosanna Nuzzi, Sergio Papa
ABSTRACT
Immunochemical and functional evidence showing the existence in the inner membrane and matrix fraction of mammalian mitochondria of serine/threonine phosphatases acting on cAMP-dependent phosphoproteins is presented. Mg(2+)-dependent Ca(2+)-inhibitable PP2C phosphatase, associated to the inner membrane, dephosphorylates the 18 kDa (NDUFS4 gene) of complex I.