Merck
  • Chemoenzymatic synthesis and lectin array characterization of a class of N-glycan clusters.
Chemoenzymatic synthesis and lectin array characterization of a class of N-glycan clusters.
Journal of the American Chemical Society (2009-11-18)
Wei Huang, Denong Wang, Masao Yamada, Lai-Xi Wang
ABSTRACT
N-Glycans are major components of many glycoproteins. These sugar moieties are frequently involved in important physiological and disease processes via their interactions with a variety of glycan-binding proteins (GBP). Clustering effect is an important feature in many glycan-lectin interactions. We describe in this paper a chemoenzymatic synthesis of novel N-glycan clusters using a tandem endoglycosidase-catalyzed transglycosylation. It was found that the internal beta-1,2-linked GlcNAc moieties in the N-glycan core, once exposed in the nonreducing terminus, was able to serve as acceptors for transglycosylation catalyzed by Endo-A and EndoM-N175A. This efficient chemoenzymatic method allows a quick extension of the sugar chains to form a class of glycan clusters in which sugar residues are all connected by native glycosidic linkages found in natural N-glycans. In addition, a discriminative enzymatic reaction at the two GlcNAc residues could be fulfilled to afford novel hybrid clusters. Lectin microarray studies revealed unusual properties in glyco-epitope expression by this panel of structurally well-defined synthetic N-glycans. These new compounds are likely valuable for functional glycomics studies to unveil new functions of both glycans and carbohydrate-binding proteins.