Merck
  • Identification and characterization of the NMYC gene product in human neuroblastoma cells by monoclonal antibodies with defined specificities.
Identification and characterization of the NMYC gene product in human neuroblastoma cells by monoclonal antibodies with defined specificities.
Proceedings of the National Academy of Sciences of the United States of America (1986-08-01)
N Ikegaki, J Bukovsky, R H Kennett
ABSTRACT
Increased N-myc (now designated NMYC in human gene nomenclature) gene expression has been detected at the transcriptional level in certain types of neoplasms. As yet, the N-myc gene product has not been identified. To detect and characterize the N-myc gene product, we have developed monoclonal antibodies against the putative N-myc gene product made in Escherichia coli as a fusion protein. The antibodies that recognize the N-myc-specific regions were selected on the basis of their reactivities to different portions of the fusion protein. These monoclonal antibodies detect a pair of closely migrating polypeptides of 60 and 63 kDa in nuclear fractions of human neuroblastoma cells. The relative levels of the polypeptides are roughly proportional to the level of N-myc transcripts present in a panel of neuroblastoma lines. These two polypeptides have a half-life of approximately equal to 35 min, and they are indistinguishable from each other by their epitopic profiles.