Merck
  • Synthesis of benzofurazan derivatization reagents for carboxylic acids and its application to analysis of fatty acids in rat plasma by high-performance liquid chromatography-electrospray ionization mass spectrometry.
Synthesis of benzofurazan derivatization reagents for carboxylic acids and its application to analysis of fatty acids in rat plasma by high-performance liquid chromatography-electrospray ionization mass spectrometry.
Biomedical chromatography : BMC (2005-06-15)
Yuhki Tsukamoto, Tomofumi Santa, Hiroshi Saimaru, Kazuhiro Imai, Takashi Funatsu
ABSTRACT
The derivatization regents for carboxylic acids, DAABD-AE (4-[2-(N,N-dimethylamino)ethylaminosulfonyl]7-(2-aminoethylamino)-2,1,3-benzoxadiazole), MePZBD-AE ([4-(4-N-methyl)piperazinosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole) and APZBD-NHMe ([4-(4-N-aminoethyl)piperazinosulfonyl]-7-methylamino-2,1,3-benzoxadiazole) were developed for electrospray ionization-mass spectrometry (ESI-MS). The derivatization reaction with fatty acids was completed at 60 degrees C within 30 min. The derivatives of fatty acids were separated on a reversed-phase column and detected with ESI-MS. The detection limits attained for fatty acids were femtomol range and the calibration curves were linear over the range from 0.1 to 100 pmol (r2 > 0.992) for DAABD-AE and MePZBD-AE. DAABD-NHMe was applied to the analysis of fatty acids in rat plasma samples.
Product Description
Supelco
4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole, for HPLC derivatization, LiChropur, ≥95.0% (HPLC)