Merck
miR-487a promotes progression of gastric cancer by targeting TIA1.
Biochimie (2018-08-26)
Xuefeng Yang, Mingda Wang, Bohao Lin, Dongjie Yao, Jin Li, Xianchun Tang, Sanhua Li, Yun Liu, Rui Xie, Shouyang Yu
ABSTRACT
Gastric cancer (GC) is one of the most common malignancies as well as the third leading cause for cancer-related death. Molecular basis of GC are essential and critical for its therapeutic treatment, but still remain poorly understood. T-cell intracellular antigen-1 (TIA1) extensively involves in cancer progression, whereas its role and regulation mechanism in GC have not been revealed. In the present study, we found that TIA-1 protein level was down-regulated in GC tissues and TIA1 inhibited proliferation and promoted apoptosis of GC cells. Then, we used bioinformatics to predict miR-487a as the upstream regulator of TIA1 and we also observed an inverse correlation between miR-487a level and TIA-1 protein level in GC tissues. Next, we demonstrated that miR-487a directly targeted TIA1 via binding to its 3'-untranslated region. Furthermore, we investigated the role of miR-487a-TIA1 pathway in the growth of GC cells both in vitro and in vivo. The repression of TIA-1 by miR-487a promoted cell proliferation and suppressed cell apoptosis in vitro, and the knockdown of miR-487a had the opposite effects. Finally, we demonstrated that miR-487a promoted the development of gastric tumor growth in xenograft mice by targeting TIA-1. These effects could be partially reversed by restoring the expression of TIA-1. Overall, our results reveal that TIA1 is a tumor suppressor gene and is directly regulated by miR-487a in GC, which may offer new therapeutic targets for GC treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human TIA1