Merck
  • Peptidome Analysis Reveals Novel Serum Biomarkers for Children with Autism Spectrum Disorder in China.
Peptidome Analysis Reveals Novel Serum Biomarkers for Children with Autism Spectrum Disorder in China.
Proteomics. Clinical applications (2018-05-14)
Juan Yang, Yanni Chen, Xiaofan Xiong, Xiaobo Zhou, Lin Han, Lei Ni, Wenjing Wang, Xiaofei Wang, Lingyu Zhao, Dongdong Shao, Chen Huang
ABSTRACT
Autism spectrum disorder (ASD) is a neurological and developmental disorder that begins early in childhood and lasts throughout one's life. Early diagnosis is essential for ASD since early treatment can enable children with ASD to make significant gains in language and social skills, but remains challenging since there are currently no specific biomarkers of ASD. This study is aimed to identify serum biomarkers for ASD. Serum of Han Chinese children with ASD (n = 68) and age-matched healthy controls (n = 80) is analyzed using magnetic bead-based separation combined with mass spectrum. Eight potential ASD serum biomarker peaks (m/z: 3886.69, 7775.12, 2381.71, 6638.63, 3319.17, 894.34, 4968.59, and 5910.53) with higher expression in ASD group are further identified as peptide regions of plasma serine protease inhibitor precursor (SERPINA5), platelet factor 4 (PF4), fatty acid binding protein 1(FABP1), apolipoprotein C-I precursor (APOC1), alpha-fetoprotein precursor (AFP), carboxypeptidase B2 (CPB2), trace amine-associated receptor 6 (TAAR6), and isoform1 of fibrinogen alpha chain precursor (FGA). The expression of identified proteins is validated by enzyme-linked immunosorbent assay (ELISA). These findings reveal the exceptional disease etiology of ASD from a serum proteomic perspective, and the identified proteins might be potential biomarkers for ASD diagnosis.
Sigma-Aldrich
Human AFP / Alpha-fetoprotein ELISA Kit, for serum, plasma, cell culture supernatants and urine