Merck
  • 1-(m-chlorophenyl)-biguanide, a potent high affinity 5-HT3 receptor agonist.
1-(m-chlorophenyl)-biguanide, a potent high affinity 5-HT3 receptor agonist.
European journal of pharmacology (1990-06-21)
G J Kilpatrick, A Butler, J Burridge, A W Oxford
ABSTRACT
1-(m-Chlorophenyl)-biguanide (mCPBG) was examined and compared with three 5-HT3 receptor agonists in three 5-HT3 receptor models. mCPBG inhibited [3H]GR67330 binding to 5-HT3 receptors with high affinity (IC50 1.5 nM). mCPBG depolarized the rat vagus nerve with an EC50 one tenth of that for 5-HT (0.05 vs. 0.46 microM); the maximum depolarization was approximately half that for 5-HT. The mCPBG depolarization was potently blocked by the selective 5-HT3 antagonist, ondansetron (pKB 8.6 +/- 0.1). In anaesthetised cats, mCPBG potently evoked the Bezold-Jarisch reflex which was blocked by low doses of ondansetron (10 micrograms/kg i.v.). It is concluded that mCPBG is a potent, high affinity 5-HT3 receptor agonist.