- Roles of Krüpel-like factor 4 in normal homeostasis, cancer and stem cells.
Roles of Krüpel-like factor 4 in normal homeostasis, cancer and stem cells.
Acta biochimica et biophysica Sinica (2008-07-08)
Paul M Evans, Chunming Liu
PMID18604447
ABSTRACT
Krüpel-like factor 4 (KLF4) is a zinc finger-type transcription factor expressed in a variety of tissues, including the epithelium of the intestine and the skin, and it plays an important role in differentiation and cell cycle arrest. Depending on the gene targeted, KLF4 can both activate and repress transcription. Moreover, in certain cellular contexts, KLF4 can function as a tumor suppressor or an oncogene. Finally, KLF4 is important in reprogramming differentiated fibroblasts into inducible pluripotent stem cells, which highly resemble embryonic stem cells. This review summarizes what is known about the diverse functions of KLF4 as well as their molecular mechanisms.
Sigma-Aldrich
KLF4-TAT human, recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE), ≥90% (HPLC), suitable for cell culture