Merck
  • Synthesis of a single 1,8-naphthalimide fluorophore as a molecular logic lab for simultaneously detecting of Fe3+, Hg2+ and Cu2.
Synthesis of a single 1,8-naphthalimide fluorophore as a molecular logic lab for simultaneously detecting of Fe3+, Hg2+ and Cu2.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2018-02-13)
Awad I Said, Nikolai I Georgiev, Vladimir B Bojinov
ABSTRACT
A novel fluorescence sensing 1,8-naphthalimide fluorophore is synthesized and investigated. The novel probe comprising two different binding moieties is capable to detect selectively Fe3+ over the other representative metal ions as well as a combination of biologically important cations such as Fe3+, Cu2+ and Hg2+ in the physiological range without an interfering effect of the pHs. Due to the remarkable fluorescence changes in the presence of Fe3+, Hg2+ and Cu2+ ions, INH and AND logic gates are executed and the system is able to act as a single output combinatorial logic circuit with three chemical inputs.
Sigma-Aldrich
4-Nitro-1,8-naphthalic anhydride, 95%