Skip to Content
Merck
Propyl disulfide ≥98%, FG

Propyl disulfide

Propyl disulfide ≥98%, FG
Synonym(s):
Dipropyl disulfide, Propyl disulfide
Linear Formula:
CH3CH2CH2SSCH2CH2CH3
CAS Number:
Molecular Weight:
150.31
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing