
Potassium citrate

Synonym(s):
Potassium citrate tribasic monohydrate, Citric acid tripotassium salt, Tripotassium citrate
Linear Formula:
HOC(COOK)(CH2COOK)2 · H2O
CAS Number:
Molecular Weight:
324.41
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing