Skip to Content
Merck
Line position and line shape standard for powder diffraction NIST® SRM® 660c, Lanthanum hexaboride powder

Line position and line shape standard for powder diffraction

Line position and line shape standard for powder diffraction NIST® SRM® 660c, Lanthanum hexaboride powder
Synonym(s):
Lanthanum hexaboride, Lanthanum boride
Linear Formula:
LaB6
CAS Number:
Molecular Weight:
203.77
EC Number:

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Quality Segment
Special Grade
Technique