Skip to Content
Merck
Iopanoic acid European Pharmacopoeia (EP) Reference Standard

Iopanoic acid

Iopanoic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(3-Amino-2,4,6-triiodobenzyl)butyric acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropanoic acid, Iodopanoic acid
Empirical Formula (Hill Notation):
C11H12I3NO2
CAS Number:
Molecular Weight:
570.93

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing