
Grubbs catalyst® M203

Synonym(s):
Tricyclohexylphosphine[1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene][3-phenyl-1H-inden-1-ylidene]ruthenium (II) dichloride
Empirical Formula (Hill Notation):
C54H67Cl2N2PRu
CAS Number:
Molecular Weight:
947.07
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing