Skip to Content
Merck
Epitestosterone certified reference material, pharmaceutical secondary standard

Epitestosterone

Epitestosterone certified reference material, pharmaceutical secondary standard
Synonym(s):
17α-Hydroxy-4-androsten-3-one, 4-Androsten-17α-ol-3-one, cis-Testosterone, Isotestosterone
Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
Showing 1-1 of 1