Skip to Content
Merck
[D-Pen2,5]-Enkephalin hydrate ≥95% (HPLC)

[D-Pen2,5]-Enkephalin hydrate

[D-Pen2,5]-Enkephalin hydrate ≥95% (HPLC)
Synonym(s):
DPDPE
Empirical Formula (Hill Notation):
C30H39N5O7S2 · xH2O
Molecular Weight:
645.79 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing