Skip to Content
Merck
Doxepin for system suitability European Pharmacopoeia (EP) Reference Standard

Doxepin for system suitability

Doxepin for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Doxepin hydrochloride, 11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenz[b,e]oxepine hydrochloride
Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1