Skip to Content
Merck
Cinnabarinic Acid ≥98% (HPLC)

Cinnabarinic Acid

Cinnabarinic Acid ≥98% (HPLC)
Synonym(s):
2-amino-3-oxo-3H-phenoxazine-1,9-dicarboxylic acid, Cinnabaric acid
Empirical Formula (Hill Notation):
C14H8N2O6
CAS Number:
Molecular Weight:
300.22

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing