Cereulide-13C6 (cyclo[-Val-1-13C-D-O-Leu-D-Ala-3-13C-O-Val-]3) solution
Synonym(s):
cyclo[-Val-1-13C-D-O-Leu-D-Ala-3-13C-O-Val-]3
Empirical Formula (Hill Notation):
13C6C51H96N6O18
Molecular Weight:
1159.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1